Thermo Scientific Chemicals

Neocuproine hemihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 153320050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Neocuproine hemihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 153320050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153320050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
34302-69-7
IUPAC Name
bis(2,9-dimethyl-1,10-phenanthroline) hydrate
Molecular Formula
C28H26N4O
InChI Key
IEBXFSLFDFHSRD-UHFFFAOYSA-N
SMILES
O.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
159°C to 164°C
Titration with HClO4
99.0 to 101.0 %
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text