Thermo Scientific Chemicals

m-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 153410050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

m-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 153410050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153410050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1711-06-4
IUPAC Name
3-methylbenzoyl chloride
Molecular Formula
C8H7ClO
InChI Key
YHOYYHYBFSYOSQ-UHFFFAOYSA-N
SMILES
CC1=CC=CC(=C1)C(Cl)=O
Appearance (Color)
Clear colorless to light brown
Titration RCOCl
>=98.5 %
Refractive index
1.5475 to 1.5495 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text