Thermo Scientific Chemicals

Thiochroman-4-one, 98%, Thermo Scientific Chemicals

Catalog number: 153480050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Thiochroman-4-one, 98%, Thermo Scientific Chemicals

Catalog number: 153480050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153480050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3528-17-4
IUPAC Name
3,4-dihydro-2H-1-benzothiopyran-4-one
Molecular Formula
C9H8OS
InChI Key
CVQSWZMJOGOPAV-UHFFFAOYSA-N
SMILES
O=C1CCSC2=CC=CC=C12
Appearance (Color)
Orange
Appearance (Form)
Low melting solid
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text