Thiochroman-4-one, 98%, Thermo Scientific Chemicals
Thiochroman-4-one, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiochroman-4-one, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Packaging:
Glass bottle
Catalog number 153480100
Price (USD)/ Each
119.00
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
119.00
Add to cart
Thiochroman-4-one, 98%, Thermo Scientific Chemicals
Catalog number153480100
Price (USD)/ Each
119.00
-
Add to cart
Chemical Identifiers
CAS3528-17-4
IUPAC Name3,4-dihydro-2H-1-benzothiopyran-4-one
Molecular FormulaC9H8OS
InChI KeyCVQSWZMJOGOPAV-UHFFFAOYSA-N
SMILESO=C1CCSC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Orange
Appearance (Form)Low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only