Thermo Scientific Chemicals

2-Amino-3-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: 153571000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Amino-3-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: 153571000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153571000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16867-03-1
IUPAC Name
2-aminopyridin-3-ol
Molecular Formula
C5H6N2O
InChI Key
BMTSZVZQNMNPCT-UHFFFAOYSA-N
SMILES
NC1=NC=CC=C1O
HPLC
>=97.5 %
Appearance (Form)
Powder or crystalline powder
Titration with HClO4
97.5 to 102.5 %
Appearance (Color)
Beige to grey to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text