Thermo Scientific Chemicals

Ethyl levulinate, 98%, Thermo Scientific Chemicals

Catalog number: 153621000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl levulinate, 98%, Thermo Scientific Chemicals

Catalog number: 153621000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153621000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
539-88-8
IUPAC Name
ethyl 4-oxopentanoate
Molecular Formula
C7H12O3
InChI Key
GMEONFUTDYJSNV-UHFFFAOYSA-N
SMILES
CCOC(=O)CCC(C)=O
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4210 to 1.4240 (20°C, 589 nm)
Density
(20°C) 1.009 to 1.014 g/ml

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text