Ethyl levulinate, 98%, Thermo Scientific Chemicals
Ethyl levulinate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl levulinate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 153625000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS539-88-8
IUPAC Nameethyl 4-oxopentanoate
Molecular FormulaC7H12O3
InChI KeyGMEONFUTDYJSNV-UHFFFAOYSA-N
SMILESCCOC(=O)CCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4210 to 1.4240 (20°C, 589 nm)
Density(20°C) 1.009 to 1.014 g/ml
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only