Thermo Scientific Chemicals

Acetamide, 99%, pure, Thermo Scientific Chemicals

Catalog number: 153630025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Acetamide, 99%, pure, Thermo Scientific Chemicals

Catalog number: 153630025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 153630025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
60-35-5
IUPAC Name
acetamide
Molecular Formula
C2H5NO
InChI Key
DLFVBJFMPXGRIB-UHFFFAOYSA-N
SMILES
CC(N)=O
Appearance (Color)
White
Appearance (Form)
Crystals
Infrared spectrum
Conforms
Melting point
78°C to 81°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text