Thermo Scientific Chemicals

Aminoacetonitrile bisulfate, 98%, Thermo Scientific Chemicals

Catalog number: 153725000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Aminoacetonitrile bisulfate, 98%, Thermo Scientific Chemicals

Catalog number: 153725000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153725000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
151-63-3
IUPAC Name
2-aminoacetonitrile; sulfuric acid
Molecular Formula
C2H6N2O4S
InChI Key
GTGIXCPOLMWQTC-UHFFFAOYSA-N
SMILES
NCC#N.OS(O)(=O)=O
Appearance (Color)
White to yellow to light beige
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text