Thermo Scientific Chemicals

2-Aminobenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: 153730250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Aminobenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: 153730250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153730250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5344-90-1
IUPAC Name
(2-aminophenyl)methanol
Molecular Formula
C7H9NO
InChI Key
VYFOAVADNIHPTR-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1CO
Appearance (Color)
Beige to grey-brown
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Melting point
82°C to 86°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text