Thermo Scientific Chemicals

2-Amino-5-bromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: 153740250
25 g, Each
Thermo Scientific Chemicals

2-Amino-5-bromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: 153740250
25 g, Each
Quantity
Catalog number: 153740250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1072-97-5
IUPAC Name
5-bromopyridin-2-amine
Molecular Formula
C5H5BrN2
InChI Key
WGOLHUGPTDEKCF-UHFFFAOYSA-N
SMILES
NC1=CC=C(Br)C=N1
Appearance (Form)
Crystalline powder
Melting point
135°C to 140°C
GC
>=96.0 %
Infrared spectrum
Conforms
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text