Thermo Scientific Chemicals

2-Aminoethanethiol hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 153775000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Aminoethanethiol hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 153775000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153775000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
156-57-0
IUPAC Name
hydrogen 2-aminoethane-1-thiol chloride
Molecular Formula
C2H8ClNS
InChI Key
OGMADIBCHLQMIP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCCS
Appearance (Color)
White
Loss on drying
=<1 % (50°C, 4 h, 100 mmHg)
Appearance (Form)
Crystalline powder or crystals and/or lumps
Melting point
66°C to 70°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text