Thermo Scientific Chemicals

2-(4-Aminophenyl)ethylamine, 95%, Thermo Scientific Chemicals

Catalog number: 153840100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(4-Aminophenyl)ethylamine, 95%, Thermo Scientific Chemicals

Catalog number: 153840100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153840100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13472-00-9
IUPAC Name
4-(2-aminoethyl)aniline
Molecular Formula
C8H12N2
InChI Key
LNPMZQXEPNWCMG-UHFFFAOYSA-N
SMILES
NCCC1=CC=C(N)C=C1
Appearance (Color)
Clear colorlesss to yellow-brown
Refractive index
1.5890 to 1.5930 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text