Thermo Scientific Chemicals

2-Aminothiophenol, 98%, Thermo Scientific Chemicals

Catalog number: 153901000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Aminothiophenol, 98%, Thermo Scientific Chemicals

Catalog number: 153901000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153901000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
137-07-5
IUPAC Name
2-aminobenzene-1-thiol
Molecular Formula
C6H7NS
InChI Key
VRVRGVPWCUEOGV-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1S
Appearance (Color)
Clear yellow to orange
Infrared spectrum
Conforms
Refractive index
1.6410 to 1.6450 (20°C, 589 nm)
Appearance (Form)
Liquid
Titration Argentometric
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text