Trypsin-EDTA (0.5%), no phenol red
Save up to 31% on your order of cell culture products. View promotion details »
Trypsin-EDTA (0.5%), no phenol red
Gibco™

Trypsin-EDTA (0.5%), no phenol red

Gibco Trypsin-EDTA is made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Due to its digestiveRead more
Have Questions?
Catalog number 15400054
Price (USD)/ 100 ml
40.65
Online exclusive
45.55 
Save 4.90 (11%)
-
Add to cart
Customize this product
Price (USD)/ 100 ml
40.65
Online exclusive
45.55 
Save 4.90 (11%)
Add to cart
Trypsin-EDTA (0.5%), no phenol red
Catalog number15400054
Price (USD)/ 100 ml
40.65
Online exclusive
45.55 
Save 4.90 (11%)
-
Add to cart
Gibco Trypsin-EDTA is made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, trypsin solution is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. Concentrated forms of trypsin can be aseptically diluted to 1X using a balanced salt solution without calcium and magnesium. The trypsin concentration required for dissociation varies with cell type and experimental requirements. See the complete range of Gibco trypsin solutions and recommended dissociation conditions.

We offer a variety of trypsin formulations and animal origin-free TrypLE reagent that feature:
• Quality testing
• Documented traceability
• Dual-site cGMP Manufacturing

This trypsin is modified as follows:

With
• EDTA

Without
• Phenol Red

The complete formulation is available.

Quality testing
Gibco Trypsin solutions are tested for pH, osmolality, sterility, and performance. In addition, the raw materials are verified for e-beam irradiation and precursor lots are tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis.

Documented traceability
We can provide detailed documentation to meet your regulatory needs. Gibco Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.

cGMP manufacturing and quality system
For supply chain continuity, we manufacture Gibco Trypsin-EDTA (0.5%), no phenol red at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
ChelatorsEDTA
Cell TypeMammalian Cells
ClassificationAnimal Origin
Concentration10 X
FormLiquid
Product TypeCell Culture Dissociation Reagent
Shelf Life24 months
SterilitySterile-filtered
With AdditivesEDTA
Without AdditivesNo Phenol Red
Green FeaturesSustainable packaging
Osmolality280 - 340 mOsm/kg
pH7.1 to 8.0
Product LineGibco™
Quantity100 mL
Shipping ConditionWet Ice
SourceAnimal Origin
Tested ForIn vitro Bioassay
Unit Size100 ml
Contents & Storage
Storage conditions: -5°C to -20°C