3,4-Dihydroxy-3-cyclobutene-1,2-dione, 99%
3,4-Dihydroxy-3-cyclobutene-1,2-dione, 99%
3,4-Dihydroxy-3-cyclobutene-1,2-dione, 99%
3,4-Dihydroxy-3-cyclobutene-1,2-dione, 99%
Thermo Scientific Chemicals

3,4-Dihydroxy-3-cyclobutene-1,2-dione, 99%

CAS: 2892-51-5 | C4H2O4 | 114.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 154020050
Price (EUR)
60,52
Online Exclusive
71,20
Save 10,68 (15%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
60,52
Online Exclusive
71,20
Save 10,68 (15%)
Each
Add to cart
Chemical Identifiers
CAS2892-51-5
IUPAC Namedihydroxycyclobut-3-ene-1,2-dione
Molecular FormulaC4H2O4
InChI KeyPWEBUXCTKOWPCW-UHFFFAOYSA-N
SMILESOC1=C(O)C(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Titration with NaOH>=98.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
I used the InVision His-Tag In-Gel Stain and am getting high, uneven background. Can you please offer some tips?
I have stained my His-tagged fusion protein with InVision His-Tag In-Gel Stain. Can I perform western blotting of the stained protein?
What is the sensitivity of the Invision His-Tag In-Gel Stain?
What is the composition of the InVision His-Tag In-Gel Stain?
Can I stain E-PAGE gels with InVision His-Tag In-Gel Stain?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471138Certificate of AnalysisMay 17, 2025154020250, 154020050, 154021000
A0461485Certificate of AnalysisApr 04, 2024154020250, 154020050, 154021000
A0397225Certificate of AnalysisDec 15, 2022154020250, 154020050, 154021000
A0404105Certificate of AnalysisDec 13, 2022154020250, 154020050, 154021000
A0405957Certificate of AnalysisDec 13, 2022154020250, 154020050, 154021000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

Share catalog number, name or link

1x1 image pixel for data collection