Thermo Scientific Chemicals

cis-1,2-Cyclohexanedicarboxylic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: 154030010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

cis-1,2-Cyclohexanedicarboxylic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: 154030010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 154030010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13149-00-3
IUPAC Name
octahydro-2-benzofuran-1,3-dione
Molecular Formula
C8H10O3
InChI Key
MUTGBJKUEZFXGO-UHFFFAOYNA-N
SMILES
O=C1OC(=O)C2CCCCC12
Appearance (Color)
White to off-white
Melting point
29°C to 32°C
Appearance (Form)
Low melting crystalline solid
Color of molten substance
=<50 APHA
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text