Thermo Scientific Chemicals

Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals

Catalog number: 154051000
100 g, Each
Thermo Scientific Chemicals

Benzenesulfinic acid, sodium salt, 97%, Thermo Scientific Chemicals

Catalog number: 154051000
100 g, Each
Quantity
Catalog number: 154051000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
873-55-2
IUPAC Name
benzenesulfinate
Molecular Formula
C6H5O2S
InChI Key
JEHKKBHWRAXMCH-UHFFFAOYSA-M
SMILES
[O-]S(=O)C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
White
Titration with HClO4
96.0 to 104.0 %
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text