Thermo Scientific Chemicals

1-Benzyl-2-pyrrolidinone, 97%, Thermo Scientific Chemicals

Catalog number: 154121000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Benzyl-2-pyrrolidinone, 97%, Thermo Scientific Chemicals

Catalog number: 154121000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 154121000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5291-77-0
IUPAC Name
1-benzylpyrrolidin-2-one
Molecular Formula
C11H13NO
InChI Key
LVUQCTGSDJLWCE-UHFFFAOYSA-N
SMILES
O=C1CCCN1CC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Refractive index
1.5500 to 1.5530 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text