2-Bromofluorene, 95%, Thermo Scientific Chemicals
2-Bromofluorene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromofluorene, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 154220250
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1133-80-8
IUPAC Name2-bromo-9H-fluorene
Molecular FormulaC13H9Br
InChI KeyFXSCJZNMWILAJO-UHFFFAOYSA-N
SMILESBrC1=CC=C2C(CC3=CC=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=94 %
Appearance (Form)Crystalline powder or flakes
Melting point110°C to 115°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only