5-Bromoisatin, 98%, Thermo Scientific Chemicals
5-Bromoisatin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromoisatin, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 154230050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS87-48-9
IUPAC Name5-bromo-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC8H4BrNO2
InChI KeyMBVCESWADCIXJN-UHFFFAOYSA-N
SMILESBrC1=CC=C2NC(=O)C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only