Thermo Scientific Chemicals

5-Bromoisatin, 98%, Thermo Scientific Chemicals

Catalog number: 154230250
25 g, Plastic Bottle, Each
Thermo Scientific Chemicals

5-Bromoisatin, 98%, Thermo Scientific Chemicals

Catalog number: 154230250
25 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 154230250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
87-48-9
IUPAC Name
5-bromo-2,3-dihydro-1H-indole-2,3-dione
Molecular Formula
C8H4BrNO2
InChI Key
MBVCESWADCIXJN-UHFFFAOYSA-N
SMILES
BrC1=CC=C2NC(=O)C(=O)C2=C1
Appearance (Color)
Orange
HPLC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text