Thermo Scientific Chemicals

1-Bromo-2-methylnaphthalene, 90%, technical, Thermo Scientific Chemicals

Catalog number: 154260250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Bromo-2-methylnaphthalene, 90%, technical, Thermo Scientific Chemicals

Catalog number: 154260250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 154260250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2586-62-1
IUPAC Name
1-bromo-2-methylnaphthalene
Molecular Formula
C11H9Br
InChI Key
CMIMBQIBIZZZHQ-UHFFFAOYSA-N
SMILES
CC1=CC=C2C=CC=CC2=C1Br
GC
>=88 %
Appearance (Form)
Liquid
Refractive index
1.6450 to 1.6530 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text