Thermo Scientific Chemicals

2-Bromothiophene, 98%, Thermo Scientific Chemicals

Catalog number: 154292500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromothiophene, 98%, Thermo Scientific Chemicals

Catalog number: 154292500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 154292500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1003-09-4
IUPAC Name
2-bromothiophene
Molecular Formula
C4H3BrS
InChI Key
TUCRZHGAIRVWTI-UHFFFAOYSA-N
SMILES
BrC1=CC=CS1
Appearance (Color)
Clear colorless to light brown
Infrared spectrum
Conforms
Refractive index
1.5850 to 1.5870 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text