1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 154315000
Price (USD)/ Each
170.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
201.00 
Save 30.35 (15%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
170.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
201.00 
Save 30.35 (15%)
Add to cart
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
Catalog number154315000
Price (USD)/ Each
170.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
201.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2425-79-8
IUPAC Name2-({4-[(oxiran-2-yl)methoxy]butoxy}methyl)oxirane
Molecular FormulaC10H18O4
InChI KeySHKUUQIDMUMQQK-UHFFFAOYNA-N
SMILESC(CCOCC1CO1)COCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4590 to 1.4650 (20°C, 589 nm)
GC>=55 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only