2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals
2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 154330025
Price (USD)/ Each
88.65
Online exclusive
98.10 
Save 9.45 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
88.65
Online exclusive
98.10 
Save 9.45 (10%)
Add to cart
2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals
Catalog number154330025
Price (USD)/ Each
88.65
Online exclusive
98.10 
Save 9.45 (10%)
-
Add to cart
Chemical Identifiers
CAS111-76-2
IUPAC Name2-butoxyethan-1-ol
Molecular FormulaC6H14O2
InChI KeyPOAOYUHQDCAZBD-UHFFFAOYSA-N
SMILESCCCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Infrared spectrumConforms
Residue after evaporation=<0.02 %
Refractive index1.4180 to 1.4220 (20°C, 589 nm)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only