Thermo Scientific Chemicals

2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 154330100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 154330100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 154330100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-76-2
IUPAC Name
2-butoxyethan-1-ol
Molecular Formula
C6H14O2
InChI Key
POAOYUHQDCAZBD-UHFFFAOYSA-N
SMILES
CCCCOCCO
Infrared spectrum
Conforms
Residue after evaporation
=<0.02 %
Refractive index
1.4180 to 1.4220 (20°C, 589 nm)
Color scale
=<10 APHA
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text