Thermo Scientific Chemicals

2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 154330250
25 L, Metal drum, Each
Thermo Scientific Chemicals

2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 154330250
25 L, Metal drum, Each
Quantity
Packaging
Catalog number: 154330250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-76-2
IUPAC Name
2-butoxyethan-1-ol
Molecular Formula
C6H14O2
InChI Key
POAOYUHQDCAZBD-UHFFFAOYSA-N
SMILES
CCCCOCCO
Appearance (Form)
Clear liquid
Water
=<0.1 % (Coulometric)
Density
(20°C) 0.899 to 0.903 kg/l
Infrared spectrum
Conforms
Residue after evaporation
=<0.02 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text