N-N-Butyldiethanolamine, 98%, Thermo Scientific Chemicals
N-N-Butyldiethanolamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-N-Butyldiethanolamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number 154360025
Price (USD)/ Each
163.65
Online exclusive
192.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
163.65
Online exclusive
192.00 
Save 28.35 (15%)
Add to cart
N-N-Butyldiethanolamine, 98%, Thermo Scientific Chemicals
Catalog number154360025
Price (USD)/ Each
163.65
Online exclusive
192.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-[butyl(2-hydroxyethyl)amino]ethan-1-ol
Molecular FormulaC8H19NO2
InChI KeyGVNHOISKXMSMPX-UHFFFAOYSA-N
SMILESCCCCN(CCO)CCO
Molecular Weight (g/mol)161.25
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.4610 to 1.4640 (20°C, 589 nm)
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only