Thermo Scientific Chemicals

Gallocyanine, Thermo Scientific Chemicals

Catalog number: 154430250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Gallocyanine, Thermo Scientific Chemicals

Catalog number: 154430250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 154430250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1562-85-2
IUPAC Name
[1-carboxy-7-(dimethylamino)-4-hydroxy-10H-phenoxazin-3-yl]oxidanylium chloride
Molecular Formula
C15H13ClN2O5
InChI Key
IQVJEUYKPJBCKY-UHFFFAOYSA-M
SMILES
[Cl-].CN(C)C1=CC=C2NC3=C(OC2=C1)C(O)=C([O+])C=C3C(O)=O
Appearance (Color)
Dark green to black
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
UV spectrum
Complies

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text