Thermo Scientific Chemicals

Phenylazoformic acid 2-phenylhydrazide, suitable for colorimetric analysis, Thermo Scientific Chemicals

Catalog number: 154450100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenylazoformic acid 2-phenylhydrazide, suitable for colorimetric analysis, Thermo Scientific Chemicals

Catalog number: 154450100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 154450100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
538-62-5
IUPAC Name
3-(phenylamino)-1-(phenylimino)urea
Molecular Formula
C13H12N4O
InChI Key
ZFWAHZCOKGWUIT-UHFFFAOYSA-N
SMILES
O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Appearance (Form)
Powder
HPLC
>=35 % (Diphenylcarbazone basis)
Appearance (Color)
Orange to brown
Infrared spectrum
Conforms
Suitability
(for Hg determination) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text