2-Coumaranone, 97%, Thermo Scientific Chemicals
2-Coumaranone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Coumaranone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number 154460050
Price (USD)/ Each
136.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
136.00
Add to cart
2-Coumaranone, 97%, Thermo Scientific Chemicals
Catalog number154460050
Price (USD)/ Each
136.00
-
Add to cart
Chemical Identifiers
IUPAC Name2,3-dihydro-1-benzofuran-2-one
Molecular FormulaC8H6O2
InChI KeyACZGCWSMSTYWDQ-UHFFFAOYSA-N
SMILESO=C1CC2=CC=CC=C2O1
Molecular Weight (g/mol)134.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light orange
Infrared spectrumConforms
GC>=96.0 %
Melting point47°C to 52°C
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only