2-Coumaranone, 97%, Thermo Scientific Chemicals
2-Coumaranone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Coumaranone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Catalog number 154460100
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS553-86-6
IUPAC Name2,3-dihydro-1-benzofuran-2-one
Molecular FormulaC8H6O2
InChI KeyACZGCWSMSTYWDQ-UHFFFAOYSA-N
SMILESO=C1CC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point47°C to 52°C
Appearance (Color)White to yellow to light orange
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only