Undecylenic acid, 98+%
Undecylenic acid, 98+%
Undecylenic acid, 98+%
Undecylenic acid, 98+%
Thermo Scientific Chemicals

Undecylenic acid, 98+%

CAS: 112-38-9 | C11H20O2 | 184.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 154492500
Price (EUR)
30,01
Online Exclusive
35,30
Save 5,29 (15%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
30,01
Online Exclusive
35,30
Save 5,29 (15%)
Each
Add to cart
Chemical Identifiers
CAS112-38-9
IUPAC Nameundec-10-enoic acid
Molecular FormulaC11H20O2
InChI KeyFRPZMMHWLSIFAZ-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=97.0 %
Appearance (Form)Low melting solid or liquid
Titration with NaOH98.0 to 102.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472863Certificate of AnalysisMay 29, 2025154495000, 154490025, 154492500
A0465157Certificate of AnalysisAug 20, 2024154495000, 154490025, 154492500
A0454993Certificate of AnalysisAug 25, 2023154495000, 154490025, 154492500
A0450766Certificate of AnalysisMay 02, 2023154495000, 154490025, 154492500
A0394569Certificate of AnalysisDec 15, 2022154495000, 154490025, 154492500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Chronic aquatic toxicity
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H412 - Harmful to aquatic life with long lasting effects

Precautionary Statements

P273 - Avoid release to the environment

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

Share catalog number, name or link

1x1 image pixel for data collection