Glycolic acid, 99%, Thermo Scientific Chemicals
Glycolic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glycolic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 154510025
Price (USD)/ Each
435.65
Online exclusive
484.00 
Save 48.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
435.65
Online exclusive
484.00 
Save 48.35 (10%)
Add to cart
Glycolic acid, 99%, Thermo Scientific Chemicals
Catalog number154510025
Price (USD)/ Each
435.65
Online exclusive
484.00 
Save 48.35 (10%)
-
Add to cart
Chemical Identifiers
CAS79-14-1
IUPAC Name2-hydroxyacetic acid
Molecular FormulaC2H4O3
InChI KeyAEMRFAOFKBGASW-UHFFFAOYSA-N
SMILESOCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystals
Water=<1 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with NaOH>=98.5 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only