Thermo Scientific Chemicals

4-Cyanobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 154550250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Cyanobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 154550250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 154550250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6068-72-0
IUPAC Name
4-cyanobenzoyl chloride
Molecular Formula
C8H4ClNO
InChI Key
USEDMAWWQDFMFY-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C(C=C1)C#N
Infrared spectrum
Conforms
Appearance (Color)
White to beige
Titration RCOCl
>=97.5 %
Appearance (Form)
Crystalline powder
Melting point
66°C to 70°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text