Thermo Scientific Chemicals

DL-Camphoroquinone, 99%, Thermo Scientific Chemicals

Catalog number: 154570100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Camphoroquinone, 99%, Thermo Scientific Chemicals

Catalog number: 154570100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 154570100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10373-78-1
IUPAC Name
1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
Molecular Formula
C10H14O2
InChI Key
VNQXSTWCDUXYEZ-UHFFFAOYNA-N
SMILES
CC1(C)C2CCC1(C)C(=O)C2=O
Appearance (Color)
Yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
199°C to 204°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text