L(-)-Carvone, 99%, Thermo Scientific Chemicals
L(-)-Carvone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L(-)-Carvone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 154590050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6485-40-1
IUPAC Name(5R)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
Molecular FormulaC10H14O
InChI KeyULDHMXUKGWMISQ-SECBINFHSA-N
SMILESCC(=C)[C@@H]1CC=C(C)C(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Specific gravity(25/25°C) 0.956 to 0.961
Infrared spectrumConforms
Specific optical rotation-57° to -62° (20°C, 589 nm) (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only