Thermo Scientific Chemicals

2-Chlorobenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 154610010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

2-Chlorobenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 154610010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 154610010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
89-98-5
IUPAC Name
2-chlorobenzaldehyde
Molecular Formula
C7H5ClO
InChI Key
FPYUJUBAXZAQNL-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1C=O
Refractive index
1.5650 to 1.5670 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text