Thermo Scientific Chemicals

1-Chlorobutane, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 154630100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

1-Chlorobutane, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 154630100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 154630100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
109-69-3
IUPAC Name
1-chlorobutane
Molecular Formula
C4H9Cl
InChI Key
VFWCMGCRMGJXDK-UHFFFAOYSA-N
SMILES
CCCCCl
Appearance (Form)
Liquid
Refractive index
1.4005 to 1.4035 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text