2-chloro-4-methylaniline, 98%, Thermo Scientific Chemicals
2-chloro-4-methylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-chloro-4-methylaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 154670250
Price (USD)/ Each
103.00
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
103.00
Add to cart
2-chloro-4-methylaniline, 98%, Thermo Scientific Chemicals
Catalog number154670250
Price (USD)/ Each
103.00
-
Add to cart
Chemical Identifiers
CAS615-65-6
IUPAC Name2-chloro-4-methylaniline
Molecular FormulaC7H8ClN
InChI KeyXGYLSRFSXKAYCR-UHFFFAOYSA-N
SMILESCC1=CC=C(N)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5740 to 1.5760 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear yellow to light brown
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only