4-Chlorothiophenol, 98%, Thermo Scientific Chemicals
4-Chlorothiophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorothiophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 154765000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-chlorobenzene-1-thiol
Molecular FormulaC6H5ClS
InChI KeyVZXOZSQDJJNBRC-UHFFFAOYSA-N
SMILESSC1=CC=C(Cl)C=C1
Molecular Weight (g/mol)144.62
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point49°C to 53°C
Appearance (Color)White to off-white
GC>=97.5 %
Appearance (Form)Crystalline solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only