Thermo Scientific Chemicals

1,4-Cyclohexanedione, 98%, Thermo Scientific Chemicals

Catalog number: 154830500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

1,4-Cyclohexanedione, 98%, Thermo Scientific Chemicals

Catalog number: 154830500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 154830500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
637-88-7
IUPAC Name
cyclohexane-1,4-dione
Molecular Formula
C6H8O2
InChI Key
DCZFGQYXRKMVFG-UHFFFAOYSA-N
SMILES
O=C1CCC(=O)CC1
Appearance (Form)
Powder or flakes
Melting point
76°C to 80°C
Appearance (Color)
Yellow to beige
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text