1,4-Cyclohexanedione, 98%, Thermo Scientific Chemicals
1,4-Cyclohexanedione, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Cyclohexanedione, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 154832500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namecyclohexane-1,4-dione
Molecular FormulaC6H8O2
InChI KeyDCZFGQYXRKMVFG-UHFFFAOYSA-N
SMILESO=C1CCC(=O)CC1
Molecular Weight (g/mol)112.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or flakes
Melting point76°C to 80°C
Infrared spectrumConforms
Appearance (Color)Yellow to beige
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only