Cyclohexene, 99%, pure, stabilized
Cyclohexene, 99%, pure, stabilized
Cyclohexene, 99%, pure, stabilized
Cyclohexene, 99%, pure, stabilized
Thermo Scientific Chemicals

Cyclohexene, 99%, pure, stabilized

CAS: 110-83-8 | C6H10 | 82.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
500 mL
Packaging:
View all
Glass bottle
Catalog number 154840010
Price (USD)
97.00
Each
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
97.00
Each
Add to cart
Chemical Identifiers
CAS110-83-8
IUPAC Namecyclohexene
Molecular FormulaC6H10
InChI KeyHGCIXCUEYOPUTN-UHFFFAOYSA-N
SMILESC1CCC=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4450 to 1.4480 (20°C, 589 nm)
StabilizerApprox. 100 ppm 2-tert-Butyl-4-methylphenol
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link