Cyclopentylacetic acid, 97%, Thermo Scientific Chemicals
Cyclopentylacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentylacetic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 154870100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1123-00-8
IUPAC Name2-cyclopentylacetic acid
Molecular FormulaC7H12O2
InChI KeyYVHAIVPPUIZFBA-UHFFFAOYSA-N
SMILESOC(=O)CC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4520 to 1.4540 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only