Cyclopropylamine, 99%, Thermo Scientific Chemicals
Cyclopropylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropylamine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 154882500
Price (USD)/ Each
439.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
517.00 
Save 77.35 (15%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
439.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
517.00 
Save 77.35 (15%)
Add to cart
Cyclopropylamine, 99%, Thermo Scientific Chemicals
Catalog number154882500
Price (USD)/ Each
439.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
517.00 
Save 77.35 (15%)
-
Add to cart
Chemical Identifiers
CAS765-30-0
IUPAC Namecyclopropanamine
Molecular FormulaC3H7N
InChI KeyHTJDQJBWANPRPF-UHFFFAOYSA-N
SMILESNC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Volatile liquid
Titration with HCl99.0 to 101.0 %
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=99 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only