Thermo Scientific Chemicals

Phthalic Anhydride, 99%, Thermo Scientific Chemicals

Catalog number: 154960010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Phthalic Anhydride, 99%, Thermo Scientific Chemicals

Catalog number: 154960010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 154960010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85-44-9
IUPAC Name
1,3-dihydro-2-benzofuran-1,3-dione
Molecular Formula
C8H4O3
InChI Key
LGRFSURHDFAFJT-UHFFFAOYSA-N
SMILES
O=C1OC(=O)C2=CC=CC=C12
Infrared spectrum
Conforms
Titration by coupling
98.5 to 101.5 %
Appearance (Form)
Flakes
Melting point
130°C to 134°C
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text