Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phthalic Anhydride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 154962500
Price (USD)/ Each
18.65
Online exclusive
22.20 
Save 3.55 (16%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
18.65
Online exclusive
22.20 
Save 3.55 (16%)
Add to cart
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
Catalog number154962500
Price (USD)/ Each
18.65
Online exclusive
22.20 
Save 3.55 (16%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,3-dihydro-2-benzofuran-1,3-dione
Molecular FormulaC8H4O3
InChI KeyLGRFSURHDFAFJT-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC=CC=C12
Molecular Weight (g/mol)148.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Flakes
Melting point130°C to 134°C
Infrared spectrumConforms
Titration by coupling98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only