Thermo Scientific Chemicals

Decyl aldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 154971000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Decyl aldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 154971000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 154971000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
112-31-2
IUPAC Name
decanal
Molecular Formula
C10H20O
InChI Key
KSMVZQYAVGTKIV-UHFFFAOYSA-N
SMILES
CCCCCCCCCC=O
Appearance (Color)
Clear colorless to light yellow
GC
>=94 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4270 to 1.4320 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text